Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELMO3
Entrez Gene ID
79767
Gene Name
engulfment and cell motility 3
Gene Aliases
CED-12, CED12, ELMO-3
Chromosome Location
Chr.16: 67199125 - 67204024 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024712.3 NP_078988.2 12 - 13 1354 82
GenBank mRNA AK023886.1 - 11 - 12 1118 82
AK056923.1 - 11 - 12 1317 82
AK300976.1 - 12 - 13 1368 82
BC015524.1 - 2 - 3 164 82
BC034410.1 - 9 - 10 966 82

View Full Product Details