The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001222.3 | NP_001213.2 | 13 - 14 | 1127 | 80 |
XM_005270199.1 | XP_005270256.1 | 14 - 15 | 1181 | 80 | |
XM_005270205.1 | XP_005270262.1 | 14 - 15 | 1181 | 80 | |
XM_017016724.1 | XP_016872213.1 | 14 - 15 | 1181 | 80 | |
XM_017016727.1 | XP_016872216.1 | 13 - 14 | 1118 | 80 | |
GenBank mRNA | L07043.1 | - | 11 - 12 | 802 | 80 |