The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080759.5 | NP_542937.3 | 3 - 4 | 1548 | 101 |
XM_011534942.2 | XP_011533244.1 | 2 - 3 | 414 | 101 | |
GenBank mRNA | AF102546.1 | - | 3 - 4 | 1548 | 101 |
AF356492.1 | - | 3 - 4 | 1548 | 101 | |
AJ005670.1 | - | 3 - 4 | 801 | 101 | |
AK001000.1 | - | 3 - 4 | 437 | 101 | |
AL079278.1 | - | 3 - 4 | 805 | 101 | |
BC021219.2 | - | 3 - 4 | 570 | 101 |