The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080759.5 | NP_542937.3 | 6 - 7 | 1998 | 85 |
XM_011534939.2 | XP_011533241.1 | 7 - 8 | 3030 | 85 | |
XM_011534942.2 | XP_011533244.1 | 5 - 6 | 864 | 85 | |
XM_017020396.1 | XP_016875885.1 | 6 - 7 | 1024 | 85 | |
GenBank mRNA | AF102546.1 | - | 6 - 7 | 1998 | 85 |
AF356492.1 | - | 6 - 7 | 1998 | 85 | |
AJ005670.1 | - | 6 - 7 | 1251 | 85 | |
AK001000.1 | - | 6 - 8 | 887 | 85 | |
AL079278.1 | - | 6 - 7 | 1255 | 85 | |
BC021219.2 | - | 6 - 7 | 1020 | 85 |