Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
Chromosome Location
Chr.16: 57882340 - 57971116 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286130.1 NP_001273059.1 26 - 27 2680 90
NM_001297.4 NP_001288.3 26 - 27 2698 90
XM_011522870.2 XP_011521172.1 14 - 15 1967 90
GenBank mRNA AF042498.1 - 26 - 27 2685 90
U58837.1 - 26 - 27 2690 90

View Full Product Details