The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135639.1 | NP_001129111.1 | 7 - 8 | 530 | 77 |
NM_001286130.1 | NP_001273059.1 | 7 - 8 | 530 | 77 | |
NM_001297.4 | NP_001288.3 | 7 - 8 | 530 | 77 | |
GenBank mRNA | AF042498.1 | - | 7 - 8 | 535 | 77 |
BQ638084.1 | - | 7 - 8 | 526 | 77 | |
U18945.1 | - | 7 - 8 | 533 | 77 | |
U58837.1 | - | 7 - 8 | 522 | 77 |