The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019589.2 | NP_062535.2 | 13 - 14 | 5802 | 136 |
XM_005267860.3 | XP_005267917.1 | 13 - 14 | 5846 | 136 | |
XM_011536966.2 | XP_011535268.1 | 14 - 15 | 4961 | 136 | |
XM_011536967.1 | XP_011535269.1 | 12 - 13 | 3728 | 136 | |
GenBank mRNA | AK090435.1 | - | 11 - 12 | 4558 | 136 |
AK297582.1 | - | 9 - 10 | 1373 | 136 | |
AL832365.1 | - | 10 - 11 | 4337 | 136 | |
BC007792.1 | - | 4 - 5 | 399 | 136 | |
L40403.1 | - | 7 - 8 | 905 | 136 |