The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164281.2 | NP_001157753.1 | 8 - 9 | 2059 | 70 |
NM_001164283.2 | NP_001157755.1 | 3 - 4 | 341 | 70 | |
NM_147200.2 | NP_671733.2 | 9 - 10 | 1850 | 70 | |
NM_147686.3 | NP_679211.2 | 8 - 9 | 2062 | 70 | |
XM_006715319.3 | XP_006715382.1 | 3 - 4 | 303 | 70 | |
XM_011535386.1 | XP_011533688.1 | 9 - 10 | 2785 | 70 | |
GenBank mRNA | AF136405.1 | - | 9 - 10 | 1758 | 70 |
AF136406.1 | - | 8 - 9 | 1849 | 70 | |
AF136407.1 | - | 8 - 9 | 1849 | 70 | |
AF272151.1 | - | 7 - 8 | 1582 | 70 | |
AF274303.1 | - | 8 - 9 | 1582 | 70 | |
AK025351.1 | - | 8 - 9 | 1656 | 70 | |
AK026602.1 | - | 7 - 8 | 1056 | 70 | |
AK314415.1 | - | 9 - 10 | 1773 | 70 | |
AL050289.1 | - | 8 - 9 | 1734 | 70 | |
BC002823.2 | - | 8 - 9 | 1645 | 70 | |
BC026723.1 | - | 3 - 4 | 260 | 70 | |
BI856094.1 | - | 3 - 4 | 339 | 70 | |
BM449775.1 | - | 5 - 6 | 524 | 70 | |
CB049654.1 | - | 3 - 4 | 308 | 70 |