Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD5
Entrez Gene ID
957
Gene Name
ectonucleoside triphosphate diphosphohydrolase 5
Gene Aliases
CD39L4, NTPDase-5, PCPH
Chromosome Location
Chr.14: 73959420 - 74019326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006720325.3 XP_006720388.1 15 - 16 1514 82
XM_017021814.1 XP_016877303.1 13 - 14 1388 82
XM_017021815.1 XP_016877304.1 16 - 17 1633 82
XM_017021817.1 XP_016877306.1 14 - 15 1373 82

View Full Product Details