The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018451.4 | NP_060921.3 | 2 - 3 | 635 | 102 |
NR_047594.1 | 2 - 3 | 635 | 102 | ||
NR_047595.1 | 2 - 3 | 635 | 102 | ||
XM_011535149.2 | XP_011533451.1 | 2 - 3 | 635 | 102 | |
XM_011535150.2 | XP_011533452.1 | 2 - 3 | 635 | 102 | |
XM_017020673.1 | XP_016876162.1 | 2 - 3 | 635 | 102 | |
GenBank mRNA | AK292406.1 | - | 2 - 3 | 635 | 102 |
AL833182.1 | - | 2 - 3 | 582 | 102 | |
BC092467.1 | - | 2 - 3 | 613 | 102 | |
BC113110.1 | - | 2 - 3 | 571 | 102 | |
BC113111.1 | - | 2 - 3 | 572 | 102 | |
BC113662.1 | - | 2 - 3 | 522 | 102 | |
BC113664.1 | - | 2 - 3 | 522 | 102 | |
BC143757.1 | - | 2 - 3 | 522 | 102 | |
BC143758.1 | - | 2 - 3 | 522 | 102 |