Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1GAP2
Entrez Gene ID
23108
Gene Name
RAP1 GTPase activating protein 2
Gene Aliases
GARNL4, RAP1GA3
Chromosome Location
Chr.17: 2755698 - 3037741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100398.1 NP_001093868.1 10 - 11 858 98
NM_015085.4 NP_055900.4 11 - 12 903 98
XM_005256542.2 XP_005256599.1 10 - 11 1295 98
XM_006721477.3 XP_006721540.1 11 - 12 844 98
XM_011523738.1 XP_011522040.1 12 - 13 956 98
XM_011523739.1 XP_011522041.1 11 - 12 911 98
XM_011523740.2 XP_011522042.1 11 - 12 816 98
XM_011523741.2 XP_011522043.1 11 - 12 992 98
XM_011523742.2 XP_011522044.1 8 - 9 698 98
XM_011523743.2 XP_011522045.1 9 - 10 776 98
XM_011523744.2 XP_011522046.1 4 - 5 364 98
XM_011523745.2 XP_011522047.1 4 - 5 321 98
XM_017024370.1 XP_016879859.1 11 - 12 881 98
XM_017024371.1 XP_016879860.1 10 - 11 934 98
GenBank mRNA AJ628446.1 - 11 - 12 903 98
AJ628447.1 - 10 - 11 858 98
AK124640.1 - 11 - 12 992 98
BC140870.1 - 11 - 12 859 98
BC140871.1 - 11 - 12 859 98
CR627427.1 - 10 - 11 827 98

View Full Product Details