Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABRD
Entrez Gene ID
2563
Gene Name
gamma-aminobutyric acid type A receptor delta subunit
Gene Aliases
EIG10, EJM7, GEFSP5
Chromosome Location
Chr.1: 2019329 - 2030753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000815.4 NP_000806.2 3 - 4 342 58
XM_011541194.2 XP_011539496.1 3 - 4 2254 58
XM_017000936.1 XP_016856425.1 2 - 3 3388 58
GenBank mRNA AF016917.1 - 3 - 4 247 58
AK290861.1 - 3 - 4 323 58
BC033801.1 - 3 - 4 258 58
BI824761.1 - 3 - 4 266 58
DA060315.1 - 3 - 4 342 58
DA064930.1 - 3 - 4 273 58

View Full Product Details