The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000815.4 | NP_000806.2 | 6 - 7 | 790 | 72 |
XM_011541194.2 | XP_011539496.1 | 6 - 7 | 2702 | 72 | |
XM_017000936.1 | XP_016856425.1 | 5 - 6 | 3836 | 72 | |
GenBank mRNA | AF016917.1 | - | 6 - 7 | 695 | 72 |
AK290861.1 | - | 6 - 7 | 771 | 72 | |
BC033801.1 | - | 6 - 7 | 706 | 72 | |
BI824761.1 | - | 6 - 7 | 72 |