Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UNC79
- Entrez Gene ID
-
57578
- Gene Name
- unc-79 homolog (C. elegans)
- Gene Aliases
-
KIAA1409
- Chromosome Location
-
Chr.14: 93333219 - 93710473 on Build GRCh38
View Full Product Details