Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOSPD2
Entrez Gene ID
158747
Gene Name
motile sperm domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 14873405 - 14922166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177475.1 NP_001170946.1 12 - 13 1402 101
NM_152581.3 NP_689794.1 13 - 14 1438 101
XM_005274451.1 XP_005274508.1 13 - 14 1438 101
GenBank mRNA AK093075.1 - 12 - 13 1402 101
AK315781.1 - 13 - 14 1378 101
AL834345.1 - 5 - 6 490 101
BC030641.1 - 13 - 14 1405 101

View Full Product Details