Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOSPD2
Entrez Gene ID
158747
Gene Name
motile sperm domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 14873405 - 14922166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177475.1 NP_001170946.1 2 - 3 318 144
NM_152581.3 NP_689794.1 3 - 4 354 144
XM_005274451.1 XP_005274508.1 3 - 4 354 144
GenBank mRNA AK093075.1 - 2 - 3 318 144
AK315781.1 - 3 - 4 295 144
BC030641.1 - 3 - 4 321 144
DA708010.1 - 3 - 4 354 144

View Full Product Details