Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAB21L3
Entrez Gene ID
126868
Gene Name
mab-21 like 3
Gene Aliases
C1orf161
Chromosome Location
Chr.1: 116072744 - 116135240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152367.2 NP_689580.2 3 - 4 458 82
XM_011540623.2 XP_011538925.1 3 - 4 590 82
XM_011540625.2 XP_011538927.1 4 - 5 686 82
XM_011540626.2 XP_011538928.1 3 - 4 512 82
XM_017000260.1 XP_016855749.1 4 - 5 1923 82
GenBank mRNA AK096035.1 - 3 - 4 458 82
BC128148.1 - 2 - 3 352 82
BC128149.1 - 2 - 3 352 82

View Full Product Details