Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCOR
Entrez Gene ID
84458
Gene Name
ligand dependent nuclear receptor corepressor
Gene Aliases
MLR2
Chromosome Location
Chr.10: 96832250 - 96964441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032440.3 NP_115816.2 1 - 2 157 84
GenBank mRNA AB058698.1 - 1 - 2 70 84
DA507116.1 - 1 - 2 157 84

View Full Product Details