The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016580.3 | NP_057664.1 | 2 - 3 | 4186 | 79 |
GenBank mRNA | AB026893.1 | - | 2 - 3 | 3032 | 79 |
AF231025.1 | - | 2 - 3 | 3281 | 79 | |
AF240635.1 | - | 2 - 3 | 4186 | 79 | |
AK023785.1 | - | 2 - 3 | 1849 | 79 | |
AK024140.1 | - | 2 - 3 | 3324 | 79 | |
AK226049.1 | - | 2 - 3 | 3283 | 79 | |
AY358428.1 | - | 2 - 3 | 3303 | 79 | |
BC042634.1 | - | 2 - 3 | 3236 | 79 | |
BC052973.1 | - | 2 - 3 | 3293 | 79 |