The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190799.1 | NP_001177728.1 | 9 - 10 | 2360 | 90 |
NM_001190801.1 | NP_001177730.1 | 10 - 11 | 2279 | 90 | |
NM_014264.4 | NP_055079.3 | 10 - 11 | 2456 | 90 | |
XM_005262701.2 | XP_005262758.1 | 10 - 11 | 2316 | 90 | |
XM_017007662.1 | XP_016863151.1 | 10 - 11 | 2615 | 90 | |
XM_017007663.1 | XP_016863152.1 | 10 - 11 | 2514 | 90 | |
GenBank mRNA | AB006972.1 | - | 10 - 11 | 2370 | 90 |
AK302858.1 | - | 10 - 11 | 2279 | 90 | |
AK303399.1 | - | 9 - 10 | 2294 | 90 | |
AK314238.1 | - | 10 - 11 | 2424 | 90 | |
BC036023.1 | - | 10 - 11 | 2424 | 90 | |
Y13115.1 | - | 10 - 11 | 2322 | 90 |