The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165257.1 | NP_001158729.1 | 3 - 4 | 749 | 151 |
NM_013243.3 | NP_037375.2 | 4 - 5 | 802 | 151 | |
GenBank mRNA | AF078851.1 | - | 4 - 5 | 684 | 151 |
AF453583.1 | - | 4 - 5 | 807 | 151 | |
AK075314.1 | - | 4 - 5 | 661 | 151 | |
AK290175.1 | - | 4 - 5 | 638 | 151 | |
AK296466.1 | - | 3 - 4 | 588 | 151 | |
AY359093.1 | - | 4 - 5 | 663 | 151 | |
BC014539.1 | - | 4 - 5 | 438 | 151 |