The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170710.4 | NP_733828.2 | 11 - 12 | 1814 | 123 |
NM_181265.3 | NP_851782.3 | 10 - 11 | 1691 | 123 | |
XM_005262725.3 | XP_005262782.1 | 9 - 10 | 1337 | 123 | |
XM_011531582.2 | XP_011529884.1 | 10 - 11 | 1679 | 123 | |
XM_017007686.1 | XP_016863175.1 | 10 - 11 | 1667 | 123 | |
XM_017007687.1 | XP_016863176.1 | 11 - 12 | 1752 | 123 | |
XM_017007688.1 | XP_016863177.1 | 11 - 12 | 1669 | 123 | |
XM_017007689.1 | XP_016863178.1 | 10 - 11 | 1666 | 123 | |
XM_017007690.1 | XP_016863179.1 | 11 - 12 | 1752 | 123 | |
XM_017007691.1 | XP_016863180.1 | 10 - 11 | 1666 | 123 | |
XM_017007692.1 | XP_016863181.1 | 11 - 12 | 1752 | 123 | |
XM_017007693.1 | XP_016863182.1 | 10 - 11 | 1666 | 123 | |
XM_017007694.1 | XP_016863183.1 | 11 - 12 | 1650 | 123 | |
XM_017007695.1 | XP_016863184.1 | 10 - 11 | 1680 | 123 | |
XM_017007696.1 | XP_016863185.1 | 10 - 11 | 1680 | 123 | |
XM_017007697.1 | XP_016863186.1 | 10 - 11 | 1668 | 123 | |
GenBank mRNA | AF492460.1 | - | 11 - 12 | 1667 | 123 |