The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170710.4 | NP_733828.2 | 14 - 15 | 2273 | 77 |
NM_181265.3 | NP_851782.3 | 13 - 14 | 2150 | 77 | |
XM_005262725.3 | XP_005262782.1 | 12 - 13 | 1796 | 77 | |
XM_011531582.2 | XP_011529884.1 | 13 - 14 | 2138 | 77 | |
XM_017007686.1 | XP_016863175.1 | 13 - 14 | 2126 | 77 | |
XM_017007687.1 | XP_016863176.1 | 14 - 15 | 2211 | 77 | |
XM_017007688.1 | XP_016863177.1 | 14 - 15 | 2128 | 77 | |
XM_017007689.1 | XP_016863178.1 | 13 - 14 | 2125 | 77 | |
XM_017007690.1 | XP_016863179.1 | 14 - 15 | 2211 | 77 | |
XM_017007691.1 | XP_016863180.1 | 12 - 13 | 1960 | 77 | |
XM_017007692.1 | XP_016863181.1 | 13 - 14 | 2046 | 77 | |
XM_017007693.1 | XP_016863182.1 | 12 - 13 | 1960 | 77 | |
XM_017007694.1 | XP_016863183.1 | 13 - 14 | 1944 | 77 | |
XM_017007695.1 | XP_016863184.1 | 12 - 13 | 1974 | 77 | |
XM_017007696.1 | XP_016863185.1 | 12 - 13 | 1974 | 77 | |
XM_017007697.1 | XP_016863186.1 | 13 - 14 | 2127 | 77 | |
GenBank mRNA | AF492460.1 | - | 14 - 15 | 2126 | 77 |