The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170710.4 | NP_733828.2 | 25 - 26 | 3527 | 103 |
NM_181265.3 | NP_851782.3 | 23 - 24 | 3359 | 103 | |
XM_005262725.3 | XP_005262782.1 | 23 - 24 | 3026 | 103 | |
XM_011531582.2 | XP_011529884.1 | 24 - 25 | 3368 | 103 | |
XM_017007686.1 | XP_016863175.1 | 24 - 25 | 3356 | 103 | |
XM_017007687.1 | XP_016863176.1 | 25 - 26 | 3441 | 103 | |
XM_017007688.1 | XP_016863177.1 | 25 - 26 | 3358 | 103 | |
XM_017007689.1 | XP_016863178.1 | 23 - 24 | 3334 | 103 | |
XM_017007690.1 | XP_016863179.1 | 24 - 25 | 3420 | 103 | |
XM_017007691.1 | XP_016863180.1 | 23 - 24 | 3190 | 103 | |
XM_017007692.1 | XP_016863181.1 | 24 - 25 | 3276 | 103 | |
XM_017007693.1 | XP_016863182.1 | 22 - 23 | 3169 | 103 | |
XM_017007694.1 | XP_016863183.1 | 23 - 24 | 3153 | 103 | |
XM_017007695.1 | XP_016863184.1 | 23 - 24 | 3204 | 103 | |
XM_017007696.1 | XP_016863185.1 | 22 - 23 | 3183 | 103 | |
GenBank mRNA | AF492460.1 | - | 25 - 26 | 3380 | 103 |
AK130128.1 | - | 9 - 10 | 989 | 103 | |
BX648508.1 | - | 9 - 10 | 927 | 103 |