Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR17
Entrez Gene ID
116966
Gene Name
WD repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.4: 176065834 - 176182828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170710.4 NP_733828.2 29 - 30 3961 157
NM_181265.3 NP_851782.3 27 - 28 3793 157
XM_005262725.3 XP_005262782.1 27 - 28 3460 157
XM_011531582.2 XP_011529884.1 28 - 29 3802 157
XM_017007686.1 XP_016863175.1 28 - 29 3790 157
XM_017007687.1 XP_016863176.1 29 - 30 3875 157
XM_017007688.1 XP_016863177.1 29 - 30 3792 157
XM_017007689.1 XP_016863178.1 27 - 28 3768 157
XM_017007690.1 XP_016863179.1 28 - 29 3854 157
XM_017007691.1 XP_016863180.1 27 - 28 3624 157
XM_017007692.1 XP_016863181.1 28 - 29 3710 157
XM_017007693.1 XP_016863182.1 26 - 27 3603 157
XM_017007694.1 XP_016863183.1 27 - 28 3587 157
XM_017007695.1 XP_016863184.1 27 - 28 3638 157
XM_017007696.1 XP_016863185.1 26 - 27 3617 157
GenBank mRNA AF492460.1 - 29 - 30 3814 157
AK130128.1 - 13 - 14 1423 157

View Full Product Details