Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR17
Entrez Gene ID
116966
Gene Name
WD repeat domain 17
Gene Aliases
-
Chromosome Location
Chr.4: 176065834 - 176182828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170710.4 NP_733828.2 2 - 3 367 145
XM_017007686.1 XP_016863175.1 1 - 2 220 145
XM_017007688.1 XP_016863177.1 2 - 3 222 145
XM_017007689.1 XP_016863178.1 1 - 2 219 145
XM_017007691.1 XP_016863180.1 1 - 2 219 145
XM_017007693.1 XP_016863182.1 1 - 2 219 145
XM_017007694.1 XP_016863183.1 2 - 3 203 145
XM_017007697.1 XP_016863186.1 1 - 2 221 145
GenBank mRNA AF492460.1 - 2 - 3 220 145

View Full Product Details