The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170710.4 | NP_733828.2 | 30 - 31 | 4156 | 144 |
NM_181265.3 | NP_851782.3 | 28 - 29 | 3988 | 144 | |
XM_005262725.3 | XP_005262782.1 | 28 - 29 | 3655 | 144 | |
XM_011531582.2 | XP_011529884.1 | 29 - 30 | 3997 | 144 | |
XM_017007686.1 | XP_016863175.1 | 29 - 30 | 3985 | 144 | |
XM_017007687.1 | XP_016863176.1 | 30 - 31 | 4070 | 144 | |
XM_017007688.1 | XP_016863177.1 | 30 - 31 | 3987 | 144 | |
XM_017007689.1 | XP_016863178.1 | 28 - 29 | 3963 | 144 | |
XM_017007690.1 | XP_016863179.1 | 29 - 30 | 4049 | 144 | |
XM_017007691.1 | XP_016863180.1 | 28 - 29 | 3819 | 144 | |
XM_017007692.1 | XP_016863181.1 | 29 - 30 | 3905 | 144 | |
XM_017007693.1 | XP_016863182.1 | 27 - 28 | 3798 | 144 | |
XM_017007694.1 | XP_016863183.1 | 28 - 29 | 3782 | 144 | |
XM_017007695.1 | XP_016863184.1 | 28 - 29 | 3833 | 144 | |
XM_017007696.1 | XP_016863185.1 | 27 - 28 | 3812 | 144 | |
GenBank mRNA | AF492460.1 | - | 30 - 31 | 4009 | 144 |
AK130128.1 | - | 14 - 15 | 1618 | 144 | |
BX648508.1 | - | 14 - 15 | 1556 | 144 |