The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198515.2 | NP_940917.1 | 8 - 9 | 993 | 122 |
XM_006717820.3 | XP_006717883.1 | 8 - 9 | 1288 | 122 | |
XM_017016206.1 | XP_016871695.1 | 8 - 9 | 911 | 122 | |
GenBank mRNA | AJ457798.1 | - | 7 - 8 | 975 | 122 |
BC044830.1 | - | 8 - 9 | 993 | 122 |