Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCA3
Entrez Gene ID
21
Gene Name
ATP binding cassette subfamily A member 3
Gene Aliases
ABC-C, ABC3, EST111653, LBM180, SMDP3
Chromosome Location
Chr.16: 2275878 - 2340746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089.2 NP_001080.2 20 - 21 3414 61
GenBank mRNA AB070929.1 - 17 - 18 2744 61
AB210044.1 - 19 - 20 3240 61
BC140895.1 - 17 - 18 2739 61
BC146866.1 - 17 - 18 2739 61
BC151840.1 - 20 - 21 3396 61
U78735.1 - 20 - 21 3261 61
X97187.1 - 19 - 20 3049 61

View Full Product Details