Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CHRNA2
- Entrez Gene ID
-
1135
- Gene Name
- cholinergic receptor nicotinic alpha 2 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 27459761 - 27479296 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U62431.1 |
- |
7 - 8 |
2173 |
92 |
View Full Product Details