The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181808.3 | NP_861524.2 | 24 - 25 | 2851 | 72 |
GenBank mRNA | AF044578.1 | - | 17 - 18 | 1355 | 72 |
AL122048.1 | - | 2 - 3 | 583 | 72 | |
AY136549.1 | - | 22 - 23 | 2454 | 72 |