The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003898.3 | NP_003889.1 | 1 - 2 | 232 | 83 |
XM_006715592.3 | XP_006715655.1 | 1 - 2 | 232 | 83 | |
XM_011536224.1 | XP_011534526.1 | 1 - 2 | 232 | 83 | |
XM_011536227.2 | XP_011534529.1 | 1 - 2 | 232 | 83 | |
XM_011536230.2 | XP_011534532.1 | 1 - 2 | 232 | 83 | |
GenBank mRNA | AF318616.1 | - | 1 - 2 | 209 | 83 |
AK294480.1 | - | 1 - 2 | 232 | 83 | |
AY152396.1 | - | 1 - 2 | 134 | 83 |