The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032951.2 | NP_116569.1 | 3 - 4 | 528 | 99 |
NM_032952.2 | NP_116570.1 | 3 - 4 | 528 | 99 | |
NM_032953.2 | NP_116571.1 | 3 - 4 | 528 | 99 | |
NM_032954.2 | NP_116572.1 | 3 - 4 | 528 | 99 | |
NR_134541.1 | 3 - 4 | 531 | 99 | ||
XM_011516277.1 | XP_011514579.1 | 3 - 4 | 510 | 99 | |
XM_011516278.1 | XP_011514580.1 | 3 - 4 | 510 | 99 | |
XM_011516279.1 | XP_011514581.1 | 3 - 4 | 510 | 99 | |
GenBank mRNA | AF156603.1 | - | 3 - 4 | 528 | 99 |
AF245470.1 | - | 3 - 4 | 480 | 99 | |
AF245471.1 | - | 3 - 4 | 480 | 99 | |
AF245472.1 | - | 3 - 4 | 480 | 99 | |
AF245473.1 | - | 3 - 4 | 480 | 99 | |
AF245474.1 | - | 3 - 4 | 480 | 99 | |
BC012925.1 | - | 3 - 4 | 502 | 99 |