The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006355.4 | NP_006346.1 | 3 - 4 | 998 | 76 |
XM_005248799.4 | XP_005248856.1 | 3 - 4 | 727 | 76 | |
XM_005248800.4 | XP_005248857.1 | 3 - 4 | 868 | 76 | |
XM_017010175.1 | XP_016865664.1 | 3 - 4 | 887 | 76 | |
GenBank mRNA | AB209906.1 | - | 2 - 3 | 840 | 76 |
AK313248.1 | - | 3 - 4 | 885 | 76 | |
BC026930.1 | - | 2 - 3 | 844 | 76 | |
BX640949.1 | - | 3 - 4 | 915 | 76 | |
CR936707.1 | - | 2 - 3 | 709 | 76 | |
U90547.1 | - | 1 - 2 | 430 | 76 |