The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152643.6 | NP_689856.6 | 11 - 12 | 1812 | 61 |
XM_017016858.1 | XP_016872347.1 | 11 - 12 | 2180 | 61 | |
XM_017016859.1 | XP_016872348.1 | 11 - 12 | 2183 | 61 | |
GenBank mRNA | AB000781.1 | - | 10 - 11 | 1708 | 61 |
AB051555.1 | - | 10 - 11 | 1906 | 61 | |
AK074179.1 | - | 1 - 2 | 78 | 61 | |
AK090457.1 | - | 10 - 11 | 2719 | 61 |