The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139248.2 | NP_640341.1 | 5 - 6 | 825 | 85 |
XM_011512530.2 | XP_011510832.1 | 6 - 7 | 1009 | 85 | |
XM_011512531.2 | XP_011510833.1 | 7 - 8 | 954 | 85 | |
XM_017005852.1 | XP_016861341.1 | 4 - 5 | 730 | 85 | |
GenBank mRNA | AK074229.1 | - | 4 - 5 | 361 | 85 |
AK122651.1 | - | 5 - 6 | 825 | 85 | |
AK298380.1 | - | 5 - 6 | 629 | 85 | |
AY036912.1 | - | 5 - 6 | 805 | 85 | |
AY093498.1 | - | 5 - 6 | 797 | 85 | |
BC064941.1 | - | 5 - 6 | 812 | 85 | |
EF186228.1 | - | 4 - 5 | 700 | 85 | |
EF186229.1 | - | 5 - 6 | 802 | 85 |