Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDEM1
Entrez Gene ID
9695
Gene Name
ER degradation enhancing alpha-mannosidase like protein 1
Gene Aliases
EDEM
Chromosome Location
Chr.3: 5187674 - 5219965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014674.2 NP_055489.1 6 - 7 1351 147
XM_011534271.2 XP_011532573.1 6 - 7 1545 147
XM_011534272.2 XP_011532574.1 7 - 8 1344 147
GenBank mRNA AK292643.1 - 6 - 7 1219 147
AK298511.1 - 6 - 7 779 147
AK302065.1 - 6 - 7 820 147
BC019088.1 - 6 - 7 1358 147
D86967.1 - 6 - 7 1277 147

View Full Product Details