Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
Chromosome Location
Chr.3: 4493348 - 4847840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099952.2 NP_001093422.2 53 - 54 7672 67
NM_001168272.1 NP_001161744.1 55 - 56 7771 67
NM_002222.5 NP_002213.5 52 - 53 7627 67
XM_005265109.2 XP_005265166.1 54 - 55 7747 67
XM_005265110.2 XP_005265167.1 53 - 54 7699 67
XM_006713131.2 XP_006713194.1 54 - 55 7750 67
XM_011533681.1 XP_011531983.1 56 - 57 7819 67
XM_011533682.2 XP_011531984.1 56 - 57 7745 67
XM_011533683.2 XP_011531985.1 56 - 57 7816 67
XM_011533684.1 XP_011531986.1 56 - 57 7792 67
XM_011533685.1 XP_011531987.1 55 - 56 7786 67
XM_011533686.1 XP_011531988.1 55 - 56 7783 67
XM_011533687.1 XP_011531989.1 55 - 56 7774 67
XM_011533688.1 XP_011531990.1 55 - 56 7747 67
XM_011533690.1 XP_011531992.1 55 - 56 7696 67
XM_017006357.1 XP_016861846.1 55 - 56 7693 67
GenBank mRNA AB208868.1 - 33 - 34 4543 67
AK294594.1 - 4 - 5 744 67
AK296235.1 - 4 - 5 768 67
AK309981.1 - 4 - 5 723 67
BC144563.1 - 14 - 15 1503 67
D26070.1 - 52 - 53 7533 67
L38019.2 - 53 - 54 7569 67
U23850.1 - 53 - 54 7421 67

View Full Product Details