The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099952.2 | NP_001093422.2 | 54 - 55 | 7769 | 107 |
NM_001168272.1 | NP_001161744.1 | 56 - 57 | 7868 | 107 | |
NM_002222.5 | NP_002213.5 | 53 - 54 | 7724 | 107 | |
XM_005265109.2 | XP_005265166.1 | 55 - 56 | 7844 | 107 | |
XM_005265110.2 | XP_005265167.1 | 54 - 55 | 7796 | 107 | |
XM_006713131.2 | XP_006713194.1 | 55 - 56 | 7847 | 107 | |
XM_011533681.1 | XP_011531983.1 | 57 - 58 | 7916 | 107 | |
XM_011533682.2 | XP_011531984.1 | 57 - 58 | 7842 | 107 | |
XM_011533683.2 | XP_011531985.1 | 57 - 58 | 7913 | 107 | |
XM_011533684.1 | XP_011531986.1 | 57 - 58 | 7889 | 107 | |
XM_011533685.1 | XP_011531987.1 | 56 - 57 | 7883 | 107 | |
XM_011533686.1 | XP_011531988.1 | 56 - 57 | 7880 | 107 | |
XM_011533687.1 | XP_011531989.1 | 56 - 57 | 7871 | 107 | |
XM_011533688.1 | XP_011531990.1 | 56 - 57 | 7844 | 107 | |
XM_011533690.1 | XP_011531992.1 | 56 - 57 | 7793 | 107 | |
XM_017006357.1 | XP_016861846.1 | 56 - 57 | 7790 | 107 | |
GenBank mRNA | AB208868.1 | - | 34 - 35 | 4640 | 107 |
AK294594.1 | - | 5 - 6 | 841 | 107 | |
AK296235.1 | - | 5 - 6 | 865 | 107 | |
AK309981.1 | - | 5 - 6 | 820 | 107 | |
BC144563.1 | - | 15 - 16 | 1600 | 107 | |
D26070.1 | - | 53 - 54 | 7630 | 107 | |
L38019.2 | - | 54 - 55 | 7666 | 107 | |
U23850.1 | - | 54 - 55 | 7518 | 107 |