Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ITPR1
- Entrez Gene ID
-
3708
- Gene Name
- inositol 1,4,5-trisphosphate receptor type 1
- Gene Aliases
-
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
- Chromosome Location
-
Chr.3: 4493348 - 4847840 on Build GRCh38
View Full Product Details