Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPR1
Entrez Gene ID
3708
Gene Name
inositol 1,4,5-trisphosphate receptor type 1
Gene Aliases
ACV, CLA4, INSP3R1, IP3R, IP3R1, PPP1R94, SCA15, SCA16, SCA29
Chromosome Location
Chr.3: 4493348 - 4847840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099952.2 NP_001093422.2 8 - 9 975 79
NM_001168272.1 NP_001161744.1 8 - 9 975 79
NM_002222.5 NP_002213.5 8 - 9 975 79
XM_005265109.2 XP_005265166.1 8 - 9 975 79
XM_005265110.2 XP_005265167.1 8 - 9 975 79
XM_006713131.2 XP_006713194.1 8 - 9 975 79
XM_011533681.1 XP_011531983.1 8 - 9 975 79
XM_011533682.2 XP_011531984.1 8 - 9 901 79
XM_011533683.2 XP_011531985.1 8 - 9 975 79
XM_011533684.1 XP_011531986.1 8 - 9 975 79
XM_011533685.1 XP_011531987.1 8 - 9 975 79
XM_011533686.1 XP_011531988.1 8 - 9 975 79
XM_011533687.1 XP_011531989.1 8 - 9 975 79
XM_011533688.1 XP_011531990.1 8 - 9 975 79
XM_011533690.1 XP_011531992.1 8 - 9 975 79
XM_011533691.1 XP_011531993.1 8 - 9 975 79
XM_011533692.2 XP_011531994.1 8 - 9 975 79
XM_017006357.1 XP_016861846.1 8 - 9 975 79
XM_017006358.1 XP_016861847.1 8 - 9 975 79
GenBank mRNA AK293752.1 - 2 - 3 161 79
AK311008.1 - 8 - 9 973 79
BC144563.1 - 8 - 9 773 79
D26070.1 - 8 - 9 881 79
L38019.2 - 8 - 9 872 79
U23850.1 - 6 - 7 625 79

View Full Product Details