Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAP2
Entrez Gene ID
23527
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Gene Aliases
CENTB2, CNT-B2
Chromosome Location
Chr.3: 195274736 - 195443176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012287.5 NP_036419.3 15 - 16 1635 130
XM_006713557.3 XP_006713620.1 16 - 17 1656 130
XM_011512602.2 XP_011510904.1 16 - 17 1656 130
XM_011512603.2 XP_011510905.1 15 - 16 1635 130
XM_011512604.2 XP_011510906.1 17 - 18 1815 130
XM_011512605.2 XP_011510907.1 16 - 17 1577 130
XM_011512606.2 XP_011510908.1 16 - 17 1558 130
XM_017006046.1 XP_016861535.1 17 - 18 1764 130
XM_017006047.1 XP_016861536.1 16 - 17 1743 130
XM_017006048.1 XP_016861537.1 17 - 18 1764 130
XM_017006049.1 XP_016861538.1 16 - 17 1743 130
GenBank mRNA AJ238248.1 - 15 - 16 1454 130
AK290369.1 - 15 - 16 1567 130
BC060767.1 - 15 - 16 1570 130
D26069.3 - 15 - 16 1404 130

View Full Product Details