Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAP2
Entrez Gene ID
23527
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Gene Aliases
CENTB2, CNT-B2
Chromosome Location
Chr.3: 195274736 - 195443176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012287.5 NP_036419.3 3 - 4 472 136
XM_006713557.3 XP_006713620.1 3 - 4 472 136
XM_011512602.2 XP_011510904.1 3 - 4 472 136
XM_011512603.2 XP_011510905.1 3 - 4 472 136
XM_011512604.2 XP_011510906.1 4 - 5 631 136
XM_011512605.2 XP_011510907.1 3 - 4 393 136
XM_011512606.2 XP_011510908.1 3 - 4 374 136
XM_017006046.1 XP_016861535.1 3 - 4 496 220
XM_017006047.1 XP_016861536.1 3 - 4 496 220
XM_017006048.1 XP_016861537.1 3 - 4 496 220
XM_017006049.1 XP_016861538.1 3 - 4 496 220
GenBank mRNA AK222635.1 - 3 - 4 427 136
AK290369.1 - 3 - 4 404 136
BC060767.1 - 3 - 4 407 136
D26069.3 - 3 - 4 241 136
DA774230.1 - 3 - 4 427 136
DB087306.1 - 3 - 4 472 136

View Full Product Details