Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLE6
Entrez Gene ID
79816
Gene Name
transducin like enhancer of split 6
Gene Aliases
GRG6, PREMBL
Chromosome Location
Chr.19: 2977410 - 2995184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143986.1 NP_001137458.1 12 - 13 1096 126
NM_024760.2 NP_079036.1 11 - 12 1018 126
XM_005259645.2 XP_005259702.1 12 - 13 1105 126
XM_011528300.2 XP_011526602.1 12 - 13 1081 126
XM_011528301.1 XP_011526603.1 8 - 9 808 126
XM_011528306.1 XP_011526608.1 8 - 9 803 126
GenBank mRNA AK024071.1 - 12 - 13 1071 126
BC007215.1 - 6 - 7 704 126
BC013986.2 - 11 - 12 976 126
BC020206.1 - 12 - 13 1053 126

View Full Product Details