Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLE6
Entrez Gene ID
79816
Gene Name
transducin like enhancer of split 6
Gene Aliases
GRG6, PREMBL
Chromosome Location
Chr.19: 2977410 - 2995184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143986.1 NP_001137458.1 3 - 4 244 60
NM_024760.2 NP_079036.1 2 - 3 166 60
XM_005259645.2 XP_005259702.1 3 - 4 253 60
XM_011528300.2 XP_011526602.1 3 - 4 229 60
GenBank mRNA AK024071.1 - 3 - 4 219 60
BC013986.2 - 2 - 3 124 60
BC020206.1 - 3 - 4 201 60
DA013757.1 - 3 - 4 254 60
DB059462.1 - 3 - 4 244 60

View Full Product Details