Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLE6
Entrez Gene ID
79816
Gene Name
transducin like enhancer of split 6
Gene Aliases
GRG6, PREMBL
Chromosome Location
Chr.19: 2977410 - 2995184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143986.1 NP_001137458.1 4 - 5 291 79
NM_024760.2 NP_079036.1 3 - 4 213 79
XM_005259645.2 XP_005259702.1 4 - 5 300 79
XM_011528300.2 XP_011526602.1 4 - 5 276 79
GenBank mRNA AK024071.1 - 4 - 5 266 79
BC013986.2 - 3 - 4 171 79
BC020206.1 - 4 - 5 248 79
DA013757.1 - 4 - 5 301 79
DB059462.1 - 4 - 5 291 79

View Full Product Details