Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLE6
Entrez Gene ID
79816
Gene Name
transducin like enhancer of split 6
Gene Aliases
GRG6, PREMBL
Chromosome Location
Chr.19: 2977410 - 2995184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143986.1 NP_001137458.1 5 - 6 335 64
NM_024760.2 NP_079036.1 4 - 5 257 64
XM_005259645.2 XP_005259702.1 5 - 6 344 64
XM_011528300.2 XP_011526602.1 5 - 6 320 64
GenBank mRNA AK024071.1 - 5 - 6 310 64
BC013986.2 - 4 - 5 215 64
BC020206.1 - 5 - 6 292 64
DA013757.1 - 5 - 6 345 64

View Full Product Details