Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLE6
Entrez Gene ID
79816
Gene Name
transducin like enhancer of split 6
Gene Aliases
GRG6, PREMBL
Chromosome Location
Chr.19: 2977410 - 2995184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143986.1 NP_001137458.1 10 - 11 809 69
NM_024760.2 NP_079036.1 9 - 10 731 69
XM_005259645.2 XP_005259702.1 10 - 11 818 69
XM_011528300.2 XP_011526602.1 10 - 11 794 69
XM_011528301.1 XP_011526603.1 6 - 7 521 69
XM_011528306.1 XP_011526608.1 6 - 7 516 69
GenBank mRNA AK024071.1 - 10 - 11 784 69
BC007215.1 - 4 - 5 417 69
BC013986.2 - 9 - 10 689 69
BC020206.1 - 10 - 11 766 69

View Full Product Details