Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC66
Entrez Gene ID
285331
Gene Name
coiled-coil domain containing 66
Gene Aliases
-
Chromosome Location
Chr.3: 56557125 - 56621837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012506.4 NP_001012524.4 11 - 12 1634 104
NM_001141947.1 NP_001135419.1 11 - 12 1653 104
NR_024460.1 11 - 12 1634 104
XM_005265081.3 XP_005265138.1 11 - 12 1653 104
XM_005265082.3 XP_005265139.1 11 - 12 1632 104
XM_005265083.3 XP_005265140.1 11 - 12 1524 104
XM_005265084.3 XP_005265141.1 11 - 12 1647 104
XM_011533614.2 XP_011531916.1 10 - 11 1658 104
XM_011533615.2 XP_011531917.1 10 - 11 1668 104
XM_011533616.2 XP_011531918.1 11 - 12 1592 104
XM_011533617.2 XP_011531919.1 12 - 13 1750 104
XM_011533619.2 XP_011531921.1 12 - 13 1653 104
XM_017006229.1 XP_016861718.1 11 - 12 1632 104
XM_017006230.1 XP_016861719.1 10 - 11 1636 104
XM_017006231.1 XP_016861720.1 10 - 11 1657 104
XM_017006232.1 XP_016861721.1 11 - 12 1524 104
XM_017006233.1 XP_016861722.1 11 - 12 1503 104
XM_017006234.1 XP_016861723.1 11 - 12 1692 104
XM_017006235.1 XP_016861724.1 12 - 13 1750 104
XM_017006236.1 XP_016861725.1 13 - 14 1745 104
XM_017006237.1 XP_016861726.1 13 - 14 1782 104
XM_017006238.1 XP_016861727.1 14 - 15 1923 104
XM_017006239.1 XP_016861728.1 8 - 9 983 104
XM_017006240.1 XP_016861729.1 12 - 13 1704 104
XM_017006241.1 XP_016861730.1 13 - 14 1663 104
XM_017006242.1 XP_016861731.1 12 - 13 1526 104
XM_017006243.1 XP_016861732.1 11 - 12 1619 104
XM_017006244.1 XP_016861733.1 12 - 13 1755 104
XM_017006245.1 XP_016861734.1 10 - 11 1440 104
GenBank mRNA AL832692.1 - 11 - 12 1573 104
AL833326.1 - 12 - 13 1668 104
BC047509.1 - 11 - 12 1602 104
BC132827.1 - 11 - 12 1620 104

View Full Product Details