Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIIP
Entrez Gene ID
60672
Gene Name
migration and invasion inhibitory protein
Gene Aliases
IIP45
Chromosome Location
Chr.1: 12019242 - 12032049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021933.3 NP_068752.2 4 - 5 940 73
XM_005263487.3 XP_005263544.1 4 - 5 769 73
XM_011541895.1 XP_011540197.1 4 - 5 771 73
XM_011541896.1 XP_011540198.1 4 - 5 664 73
GenBank mRNA AK024020.1 - 4 - 5 659 73
AK095928.1 - 1 - 2 54 73
BC008068.2 - 4 - 5 649 73
BI828925.1 - 4 - 5 697 73
FJ618905.1 - 4 - 5 940 73

View Full Product Details